missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution pH: 1.679, Fisher Chemical™ Product Code.: 12910654
compliance-icons

Buffer Solution pH: 1.679, Fisher Chemical™
BIOPHARMA

Product Code. 12910654
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 12910654

Brand: Fisher Chemical SOL 007 500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

pH 1.679
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Formulation Tetraoxalate Potassium
compliance-icons
Product Identifier
  • Buffer solution - pH 1.679 @ 25°C Potassium Tetraoxalate 0.05M (SOL 007)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH: 1.679, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.