missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 1.679, Reagecon™
BIOPHARMA

Reagecon's pH Buffer Solution at 25°C is tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable, has guaranteed stability throughout its entire shelf life, even after opening the bottle.

31.82 € - 57.85 €

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 1.679 ±0.01
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11862422
View Documents Promotion Details
Reagecon
42508
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11832643
BIOPHARMA
View Documents Promotion Details
Reagecon
101679
500 mL
57.85 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Calibrated to a standard precision NIST value
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
Yes
100°C
1.679 ±0.01
Characteristic
Fully miscible in water
Videos
SDS
Documents

Documents

Product Certifications