missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide, Honeywell Fluka™
Borax / sodium hydroxide
21.11 € - 327.88 €
Chemical Identifiers
CAS | 1310-73-2, 1330-43-4, 7732-18-5 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15694040
|
Honeywell-Fluka
33649-100ML |
100 mL | N/A | ||||||
15614050
|
Honeywell-Fluka
33649-500ML |
500 mL |
91.98 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624050
|
Honeywell-Fluka
33649-6X1L |
6 x 1 L |
327.88 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1310-73-2, 1330-43-4, 7732-18-5 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
CHEBI:18219 |
H5NO | |
MFCD00066650 | |
14923 | |
N.O |
Specifications
1310-73-2, 1330-43-4, 7732-18-5 | |
H5NO | |
NONH for all modes of transport | |
N.O | |
35.05 | |
CHEBI:18219 |
10.0 | |
MFCD00066650 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
Buffer solution |
Safety and Handling
Hazard Statement
Precautionary Statement
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.