missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
pH | 10.0 |
Physical Form | Liquid |
Quantity | 1000 mL |
Solution Type | Buffer |
Formulation | Ammonium Chloride |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1050636 | Certificate of Analysis | 03/03/2025 | SOL 100 1L |
Safety and Handling


Product Identifier
- Ammonium chloride,Buffer solution - pH 10.0 European Pharmacopoeia Ref: 4007300 (SOL 100)
- Danger
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution pH: 10.0, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction