missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, Reagecon™
BIOPHARMA

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 10.00 is tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 10.0
Physical Form Liquid
View More Specs

Products 3
Product Code Brand Quantity pH Price Quantity & Availability  
Product Code Brand Quantity pH Price Quantity & Availability  
11422024
BIOPHARMA
View Documents Promotion Details
Reagecon
11005
500 mL 10.0 N/A
N/A
11496424
BIOPHARMA
View Documents Promotion Details
Reagecon
1100525
500 mL 10.0 N/A
N/A
Edge
11412024
BIOPHARMA
View Documents Promotion Details
Reagecon
1100
1000 mL 10.0 N/A
N/A
Edge
Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
Characteristic
Fully miscible in water
100°C
10.0
pH Buffer
Yes
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 a 20°C

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : boric acid, disodiumsalt

SDS
Documents

Documents

Product Certifications