missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10, Thermo Scientific Chemicals
34.30 € - 111.00 €
Specifications
Chemical Name or Material | Buffer solution pH 10 |
---|---|
Color | Colorless |
CAS | 1332-57-6, 144-55-8, 7732-18-5 |
pH | 9.98 to 10.02 (20°C) |
Physical Form | Liquid |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031891
|
Thermo Scientific Acros
258605000 |
500 mL | Plastic bottle |
34.30 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10359240
|
Thermo Scientific Acros
258600010 |
1 L | Plastic bottle |
54.10 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11916971
|
Thermo Scientific Acros
258600025 |
2.5 L | Plastic bottle |
111.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferSpecifications
Buffer solution pH 10 | |
1332-57-6, 144-55-8, 7732-18-5 | |
Liquid |
Colorless | |
9.98 to 10.02 (20°C) | |
Plastic bottle |
Safety and Handling
Product Identifier
- Buffer solution pH 10, Carbonate buffer
RUO – Research Use Only