missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 11.00, Reagecon™
BIOPHARMA

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 11.00 is tested at 20°C to an accuracy level of ±0.05. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

48.10 € - 63.45 €

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 11.00 ±0.05
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11464529
BIOPHARMA
View Documents Promotion Details
Reagecon
11105
500 mL
48.10 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11432024
BIOPHARMA
View Documents Promotion Details
Reagecon
1110
1000 mL
63.45 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
Characteristic
Fully miscible in water
100°C
11.00 ±0.05
pH Buffer
Yes
Videos
SDS
Documents

Documents

Product Certifications