missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Ph 2.0 (20 °C), Honeywell Fluka™
Citric acid / hydrochloric acid / sodium chloride, with fungicide, traceable to SRM from NIST and PTB
35.44 € - 361.26 €
Specifications
Name Note | With fungicide |
---|---|
CAS | 7732-18-5, 7647-14-5, 77-92-9 |
MDL Number | MFCD00146206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633180
|
Honeywell-Fluka
31045-100ML |
100 mL |
35.44 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643180
|
Honeywell-Fluka
31045-1L |
1 L |
69.33 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653180
|
Honeywell Fluka
31045-6X1L |
6 x 1 L |
361.26 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
With fungicide | |
7732-18-5, 7647-14-5, 77-92-9 | |
MFCD00146206 |
Safety and Handling
H290
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Buffer solution pH 2.0 (20 °C)
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Citric acid, Sodiumchloride, hydrochloric acid