missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™

Product Code. 10703324
Click to view available options
Quantity:
500 mL
1 L
2.5 L

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 217672

Specifications

Color Colorless
pH 2
Physical Form Liquid
Quantity 500 mL
Packaging HDPE plastic bottle
compliance-icons
Product Identifier
  • Buffer solution pH 2 (glycine) Standard Buffer Solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'