missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 2, Reagecon™
BIOPHARMA

40.10 € - 69.70 €

Specifications

pH 2

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11848773
BIOPHARMA
View Documents
Reagecon
1020525
500 mL
69.70 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11464539
BIOPHARMA
View Documents
Reagecon
102025
1 L
40.10 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.