missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4, Citrate buffer, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 258580010
46.77 EUR valid until 2024-12-31
Use promo code "21980" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferSpecifications
Buffer solution pH 4 | |
68-04-2, 7647-14-5, 7732-18-5 | |
3.98 to 4.02 (20°C) | |
1 L | |
MFCD00130806 | |
Traceable to NIST |
Phthalate buffer | |
Colorless | |
Liquid | |
C8H5KO4 | |
Plastic bottle | |
204.23 |
RUO – Research Use Only