missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4, Citrate buffer

Product Code. p-215503 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
5 L
missing translation for 'unitSize'
1 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 10661832

missing translation for 'mfr': Thermo Scientific Acros 258585000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-04-2 | C8H5KO4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 4
Name Note Phthalate buffer
CAS 68-04-2, 7647-14-5, 7732-18-5
Color Colorless
pH 3.98 to 4.02 (20°C)
Physical Form Liquid
Quantity 500 mL
Molecular Formula C8H5KO4
MDL Number MFCD00130806
Packaging Plastic bottle
Traceability to NIST Traceable to NIST
Solubility Information Solubility in water: soluble.
Formula Weight 204.23
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.