missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 5.00, Reagecon™
33.20 € - 52.50 €
Specifications
pH | 5.00 |
---|---|
Physical Form | Liquid |
Solution Type | Buffer |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432014
|
Reagecon
10505 |
500 mL |
33.20 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440459
|
Reagecon
1050 |
1 L |
52.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
5.00 | |
Liquid | |
Buffer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title