missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 7.00, Yellow, Reagecon™
Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 7.00, Yellow is tested at 20°C to an accuracy of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, is color coded and is conveniently packaged.
Brand: Reagecon BPH03
This item is not returnable.
View return policy
Description
Color coded and packaged for convenience
- Colored yellow for easy identification of pH value
- Supplied in Bag in Box bulk cubitainer with integrated tap for easy dispensing or Twin Neck bottles that feature an integrated calibrating chamber
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
Yellow | |
Liquid | |
Characteristic | |
Yes |
7.00 ±0.01 | |
5 L | |
Bag in Box cubitainer with integrated tap | |
Fully miscible in water |