missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 7.4 EU Pharmacopoeia, Fisher Chemical™

Product Code. 12920724
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 1336-21-6
pH 7.4
Quantity 1000 mL
Molecular Formula H5NO
MDL Number MFCD00066650
Packaging Bottle
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Grade Eu. Ph. Reference
Chemical Name or Material Buffer solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.