missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 7, Phosphate buffer, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 258590025

 View more versions of this product

Product Code. 11906971

  • 76.28 € / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer solution pH 7
1.0030g/mL
H2KO4P
Plastic bottle
14, 7659
Solubility in water: soluble.
7558-79-4, 7732-18-5
2.5 L
MFCD03097944
1.003
14mmHg at 20°C
136.09
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer solution pH 7, Phosphate buffer

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only