missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 8.00, Reagecon™
BIOPHARMA

Product Code. 11822172
Click to view available options
Quantity:
500 mL
1 L
pH:
8
This item is not returnable. View return policy
This item is not returnable. View return policy

Certified by an ISO 17025 accredited test method and NIST traceable. Reagecon™ Buffer Solution, pH 8.00 is available in solutions tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 8
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Traceability to NIST Yes
Solubility Information Fully miscible in water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'