missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

30.00 € - 75.55 €

Specifications

CAS 877-24-7
Color Colorless
pH 5
Physical Form Liquid
MDL Number 13070
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10609483
View Documents
Fisher Chemical
J/2830/08
500 mL
30.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10588230
View Documents
Fisher Chemical
J/2830/15
1 L
53.10 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10294430
View Documents
Fisher Chemical
J/2830/17
2.5 L
75.55 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement

Specifications

Specifications

877-24-7
5
13070
Colorless
Liquid
HDPE plastic bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.