missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

43.10 € - 560.00 €

Specifications

Color Colorless
pH 8
Physical Form Liquid
MDL Number 217672
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10593854
View Documents
Fisher Chemical
J/2860/08
500 mL HDPE plastic bottle
50.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10070210
View Documents
Fisher Chemical
J/2860/15
1 L HDPE plastic bottle
43.10 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10141620
View Documents
Fisher Chemical
J/2860/17
2.5 L HDPE plastic bottle
88.90 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10164373
View Documents
Fisher Chemical
J/2860/24
10 L Bag in box
560.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement

Specifications

Specifications

Colorless
8
Liquid
217672
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications