missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer Solutions for NucleoMag™ RNA

Additional buffer solutions (e.g., lysis buffer MR1, wash buffer MR3) for use in magnetic bead-based isolation of RNA from cell and tissue samples using the NucleoMag™ RNA kit.

90.85 € - 104.00 €

Specifications

Color Colorless
For Use With (Equipment) NucleoMag™ RNA kit
CAS 593-84-0
Content And Storage Room Temperature (18°C to 25°C)
Form Liquid
View More Specs

Products 2
Product Code Brand Product Type Quantity Price Quantity & Availability  
Product Code Brand Product Type Quantity Price Quantity & Availability  
15879147
View Documents
Macherey-Nagel™
744351.125
Lysis Buffer MR1 125 mL
90.85 €
125mL
Please to purchase this item. Need a web account? Register with us today!
15889147
View Documents
Macherey-Nagel™
744353.500
Wash Buffer MR3 320 mL
104.00 €
320mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NucleoMag™ RNA

  • One-tube procedure minimizes risk of cross-contamination
  • Proven 96-well plate format
  • Small elution volumes ≥ 50 μL
  • Suitable for manual and automated processing
  • Recombinant DNase included
  • Reducing agent TCEP included (no β-mercaptoethanol necessary)
Specifications

Specifications

Colorless
593-84-0
Liquid
NucleoMag™ RNA kit
Room Temperature (18°C to 25°C)
Isolation of RNA from tissue and cell samples
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : At least 1 year

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.