missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Brand: Fisher Chemical B/4770/77
18.81 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Specifications
White | |
Solid | |
Borates, Sodium Salts, Anhydrous, Sodium Chloride, Tetra | |
Glass Jar |
9.2 | |
50 Tablets | |
81857 | |
Soluble in Water |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Buffer tablets, pH 9.2 (borate)
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Borates, tetra, sodium salts, anhydrous,Sodium chloride