Learn More
Cytiva Buffer Valve
Brand:  Cytiva 29023895
Product Code. 15289764
  • 456.00 € / Each
Explore more special offers
Description
Description
For use with the AKTA start automated protein purification system. The buffer valve is used as a switching valve for gradient formation. It enables the use of two buffers, A and B, which are required for forming gradient during runs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used as a switching valve for gradient formation
AKTA start Automated Protein Purification System