missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bunsen Burner with Tétine Connector without Pilot Light

Reaches temperatures of about 900°C with butane nozzles and about 1000°C on natural gas spouts.

Brand:  Fisherbrand™ 4006/1BIO

 View more versions of this product

Product Code. 11789767

  • 24.30 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Chrome steel burner
  • Cast iron base
  • Gas regulation by needle valve
  • For use with natural gas or butane/propane

Specifications

Specifications

Bunsen Burner
Consumption of 144L/hr.
13 mm
145 mm
1 Ea.
Butane, Propane, LPG
13 mm
Tétine Connector
T-Tine Connector
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.