Learn More
Kartell™ Burette Clamp
Kartell™ Burette Clamp used a spring loaded center arm to hold the burette firmly against the two outer arms and is protected by rubber grips.
Brand: Kartell™ 0013900
This item is not returnable.
View return policy
Description
Kartell™ Burette Clamp used a spring loaded center arm to hold the burette firmly against the two outer arms and is protected by rubber grips.
- Can fit on to support rods having diameters 8 - 14mm
- Stainless steel spring loaded centre arm holds burette firmly against the two outer arms and is protected by rubber grips
- Graduations and meniscus are visible at all times
- Suitable for all burettes
Specifications
White | |
Polypropylene | |
(1) Clamp | |
8 to 14 mm |
8-14 mm Rods | |
1 Each | |
Buret Clamp |