Learn More
BRAND™ BLAUBRAND™ Compact Automatic Burette with Glass Bottle
Fast to dismantle and easy to clean. BRAND™ BLAUBRAND™ Compact Automatic Burette with Bottle are also simple to repair because all individual components replaceable.
Brand:  BRAND™ 23921
 View more versions of this product
Product Code. 10719345
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Borosilicate glass construction
  • Class AS, DE-M marking
  • Graduations in blue
  • Detachable PTFE stopcock.
  • DIN EN ISO 385
  • Calibrated to deliver (TD, Ex).
  • All BLAUBRAND™ automatic burettes are supplied with a batch number and an accompanying batch certificate in the original packaging. On request, they are available also with an individual certificate, USP individual certificate or DAkkS calibration certificate.
  • Includes Burette tube with Schellbach stripe and automatic zeroing, stopcock with precision tip, filling tube (PVC, transparent), pumphead and 2000mL soda-lime glass bottle
  • Supports, burette clamps and rubber bellows sold separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
Borosilicate Glass
Glass Bottle
±0.05 mL
Class AS
790 mm
0.1 mL
PTFE
Automatic Buret
DIN EN ISO 385, USP