missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Pipette Cleaning and Storage Container

Polyethylene cleaning and storage container available in 10L or 12.5L is ideal to use with pipettes.

138.78 € - 157.00 €

Specifications

Diameter (Metric) 155 mm
Material Polyethylene
For Use With (Equipment) Pipette
For Use With (Application) Laboratory Use
Shape Round
View More Specs

Products 2
Product Code Brand Height (Metric) Size Price Quantity & Availability  
Product Code Brand Height (Metric) Size Price Quantity & Availability  
11857253
View Documents
Buerkle™
9650-0651
550 mm 10 L
157.00 €
Each
In Stock
to see stock.
Add to basket
 
11961501
View Documents
Buerkle™
9650-0652
660 mm 12.5 L
138.78 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
Description

Description

  • PE
  • Ideal with pipettes
Specifications

Specifications

155 mm
Pipette
Round
Polyethylene
Laboratory Use
Cleaning and storage container
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Pipette Cleaning and Storage Container

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.