Learn More
Butyl butyrate, 98%, Thermo Scientific Chemicals
19.50 € - 83.52 €
Chemical Identifiers
CAS | 109-21-7 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.21 |
MDL Number | MFCD00009450 |
InChI Key | XUPYJHCZDLZNFP-UHFFFAOYSA-N |
Synonym | butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate |
PubChem CID | 7983 |
ChEBI | CHEBI:87429 |
IUPAC Name | butyl butanoate |
SMILES | CCCCOC(=O)CCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10421481
|
Thermo Scientific Acros
259590050 |
5 mL | Glass bottle |
19.50 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10760751
|
Thermo Scientific Acros
259591000 |
100 mL | Glass bottle |
22.91 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697421
|
Thermo Scientific Acros
259590010 |
1 L | Glass bottle |
83.52 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-21-7 | |
144.21 | |
XUPYJHCZDLZNFP-UHFFFAOYSA-N | |
7983 | |
butyl butanoate |
C8H16O2 | |
MFCD00009450 | |
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate | |
CHEBI:87429 | |
CCCCOC(=O)CCC |
Specifications
1mg KOH/g max. | |
-92.0°C | |
0.8675g/mL | |
49°C | |
97.5% min. (GC) | |
CH3CH2CH2CO2(CH2)3CH3 | |
MFCD00009450 | |
15, 1558 | |
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate | |
XUPYJHCZDLZNFP-UHFFFAOYSA-N | |
butyl butanoate | |
7983 | |
144.2 | |
Liquid |
109-21-7 | |
Colorless to Yellow | |
164.0°C to 165.0°C | |
Authentic | |
C8H16O2 | |
1.4050 to 1.4070 | |
02, 271 | |
0.8675 | |
Solubility in water: 0.05%. Other solubilities: soluble in alcohol and ether | |
CCCCOC(=O)CCC | |
144.21 | |
CHEBI:87429 | |
98% | |
Butyl butyrate, 98% |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-656-8
missing translation for 'rtecsNumber' : ES8120000
missing translation for 'tsca' : TSCA
- Butyl butyrate
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.