Learn More
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
36.28 € - 629.13 €
Chemical Identifiers
CAS | 25013-16-5 |
---|---|
Molecular Formula | C11H16O2 |
Molecular Weight (g/mol) | 180.24 |
MDL Number | MFCD01779059 |
InChI Key | MRBKEAMVRSLQPH-UHFFFAOYSA-N |
Synonym | 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy |
PubChem CID | 8456 |
ChEBI | CHEBI:76358 |
IUPAC Name | 2-tert-butyl-4-methoxyphenol |
SMILES | CC(C)(C)C1=C(C=CC(=C1)OC)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10551291
|
Thermo Scientific Acros
235231000 |
100 g | Plastic bottle |
36.28 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10665282
|
Thermo Scientific Acros
235235000 |
500 g | Plastic bottle |
133.67 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10648324
|
Thermo Scientific
235230025 |
2.5 kg | Plastic bottle |
629.13 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25013-16-5 | |
180.24 | |
MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
8456 | |
2-tert-butyl-4-methoxyphenol |
C11H16O2 | |
MFCD01779059 | |
3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
CHEBI:76358 | |
CC(C)(C)C1=C(C=CC(=C1)OC)O |
Specifications
25013-16-5 | |
White to Yellow | |
113°C | |
95% min. (GC) | |
MFCD01779059 | |
3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
2-tert-butyl-4-methoxyphenol | |
8456 | |
180.24 | |
Crystalline Flakes or Crystals |
48°C to 55°C | |
268°C | |
Authentic | |
C11H16O2 | |
15,1549 | |
Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,propylene glycol,,chlorogorm,ether and acetone,slightly soluble in benzene | |
CC(C)(C)C1=C(C=CC(=C1)OC)O | |
180.24 | |
CHEBI:76358 | |
96% | |
Butylated hydroxyanisole, 96% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED:
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-563-8
missing translation for 'rtecsNumber' : SL1945000
missing translation for 'tsca' : TSCA
- Butylated hydroxyanisole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only