missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylated Hydroxytoluene, 99.5%, MP Biomedicals™
Click to view available options
Quantity:
500 g
Specifications
Specifications
Density | 1.048g/mL at 20°C |
Assay Percent Range | 99.5% |
Quantity | 500 g |
Formula Weight | 220.36 |
Physical Form | Crystalline |
Chemical Name or Material | Butylated hydroxytoluene |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Acute toxicity Category 4
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 2,6-di-tert-butyl-p-cresol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction