Learn More
Jaytec Glass™ Butyrometers Tube
Brand:  Jaytec Glass™ MP.676
Product Code. 12308965
  • 42.80 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured to BS 696.
For testing fat content in dairy products, using the Gerber method.
All products are for research use only unless otherwise indicated.
Specifications
Specifications
10.8 mL