missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture
BVC Fluid Aspiration Systems for Cell Culture provide outstanding levels of comfort and safety for working with biological and biohazardous materials.
1125.00 € - 2760.00 €
Specifications
Temperature (Metric) Storage | -10°C to +60°C |
---|---|
Temperature (English) Max. | 104°F |
Temperature (Metric) Max | 40°C |
Temperature (Metric) Min. | 10°C |
Product Code | Brand | Overall Height (Metric) | Overall Length (Metric) | Model | Plug Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Overall Height (Metric) | Overall Length (Metric) | Model | Plug Type | Price | Quantity & Availability | ||
12653315
|
BrandTech™ VACUUBRAND™
20727000 |
49 cm | 25 cm | BVC Basic |
1125.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
17728674
|
BrandTech™ VACUUBRAND™
20727302 |
43 cm | 40.8 cm | BVC Control G | UK |
2255.00 €
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
17490144
|
BrandTech™ VACUUBRAND™
20727502 |
43 cm | 40.8 cm | BVC Professional G | UK |
2760.00 €
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||
12603325
|
BrandTech™ VACUUBRAND™
20727500 |
43 cm | 40.8 cm | BVC Professional G | CEE |
2760.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
12673315
|
BrandTech™ VACUUBRAND™
20727100 |
43 cm | 23 cm | BVC Basic G |
1250.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
Description
Basic, Control and Professional models to serve your fluid aspiration needs.
- 2 or 4 L vessels for safe collection and disposal of your biological waste
- Autoclavable hydrophobic 0.2 μm biofilter protects pump or other vacuum supply from biohazardous aerosols
- The VacuuHandControl (VHC) keeps all fluid within aspiration tubing for the ultimate in containment and cleaning convenience
- G models feature a 2 L glass collection vessel with scratch and fragment protection coating and true bleach resistance
- Standard versions include a 4 L heavy duty polypropylene vacuum bottle for increased capacity
- Control and Professional models feature powerful VACUUBRAND™ chemistry design vacuum pumps with exceptionally long service life and enough power for two users
- Streamlined design incorporates bottle, controls and pump (Control and Professional models), in a lab friendly small footprint
- Quick connectors are supplied with the standard (non-G) Professional, but can be added to the other five models
BVC Basic
- The unpowered version, suitable for use with a VACUU LAN vacuum network, central vacuum system or shared pump
- Provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached
- Supplied with a stable and easy-to-clean powder coated metal stand, 0.2 μm aerosol filter, and VacuuHandControl
- Available in heavy duty polypropylene, or bleach resistant glass versions
BVC Control
- The BVC Control features a powerful whisper-quiet VACUUBRAND chemistry-design vacuum pump, eliminating the need for central vacuum supply
- Potentially biohazardous materials can be contained in a local area
- The pump is turned on and off automatically on demand when using the VacuuHandControl which further reduces the noise level
- An easy to clean touch panel adjusts and indicates the vacuum level switchpoint
- Available with either a 4 L polypropylene, or 2 L bleach resistant glass bottle
BVC Professional
- Includes an external liquid level sensor, which operates without contacting bottle contents
- Special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl
- The 4 L polypropylene bottle version is supplied with quick connectors as standard
- The 2 L glass bottle 'bleach proof' version includes hose barb connections; optional quick connectors are available
Available Accessories:
- Replacement filter 638266
- Replacement VHCpro tubing (sold per meter, minimum order 3 m) 636156
- VHCpro hand controller (one supplied with unit) 688061
- VHCpro stand 635680
- Pipette tip ejector for VHCpro 635638
- 8-channel adapter with tip ejector for VHCpro 635679
- BVC shuttle (wheeled cart to facilitate access to BVC when under a BSC) 696880
- Quick connect for second user (between VHCpro and bottle) 635807
- Standard connect for second user 699943
- Quick connect (between bottle and pump) 635808
- Replacement 4 L PP bottle (with cap) 635810
- Replacement 2 L glass bottle (with cap) 635809
- 2 L glass bottle adapter 635839
Specifications
-10°C to +60°C | |
104°F | |
40°C | |
10°C |