missing translation for 'onlineSavingsMsg'
Learn More

BW-A 78U, MedChemExpress

MedChemExpress BW-A 78U is a PDE4 inhibitor with an IC50 of 3 μM.

Encompass_Preferred

POA - 209.25 €

Specifications

Chemical Name or Material BW-A 78U
CAS 101155-02-6
Color White
Purity Grade Notes Research
Molecular Formula C13H12FN5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16420509
Encompass_Preferred
View Documents
Medchem Express
HY-100118
1 mg
POA
1mg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16405527
Encompass_Preferred
View Documents
Medchem Express
HY-100118
5 mg
209.25 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BW-A 78U is a PDE4 inhibitor with an IC50 of 3 μM.
Specifications

Specifications

BW-A 78U
White
C13H12FN5
CNC1=C2N=CN(CC3=CC=CC=C3F)C2=NC=N1
257.27
Research
Neuroscience-Neuromodulation
101155-02-6
Research
DMSO : 150 mg/mL (583.05 mM; Need ultrasonic)
257.27
99.67%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
BW-A 78U, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.