Learn More
Cytiva C 10/10 Column
Economical empty columns suitable for low pressure liquid chromatography, and gravity feed at lower volumes typically used at research scale.
Brand:  Cytiva 19-5035-01
Product Code. 10221110
  • 116.00 € / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
10 mm
Low Pressure Liquid Chromatography
Polyethylene Tubing (1 mm I.D.), Bed Support Screens, Snap-On Net Rings, and Instructions
10 mm