Learn More
Saint Gobain Life Sciences™ C-Flex™ 082 TPE Tubing
GREENER_CHOICE
Biopharmaceutical translucent C-Flex™ 082 TPE Tubing.
66.15 € - 2375.00 €
Specifications
Autoclavable Autoclavable
Material Thermoplastic
Product Type Tubing
View More Specs
Products 37
Product Code Brand Diameter (Metric) Outer Maximum Pressure Overall Length (Metric) Thickness (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Maximum Pressure Overall Length (Metric) Thickness (Metric) Price Quantity & Availability  
15650685
View Documents
Saint Gobain Life Sciences™
082-125-2
6.4 mm 1106 psi 15.2 m 1.6 mm
95.68 €
15m
In Stock
to see stock.
Add to basket
15660685
View Documents
Saint Gobain Life Sciences™
082-125-2M
6.4 mm 1106 psi 152.4 m 1.6 mm
590.00 €
152m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15670685
View Documents
Saint Gobain Life Sciences™
082-125-2S
6.4 mm 1106 psi 762.0 m 1.6 mm
2375.00 €
762m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
17973421
View Documents
Saint Gobain Life Sciences™
082-188-2M
8 mm 22 psi (at 19°C) 107 m 1.6 mm
355.00 €
107m
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15690685
View Documents
Saint Gobain Life Sciences™
082-188-2M
8 mm 1106 psi 106.7 m 1.6 mm N/A N/A N/A
15600695
View Documents
Saint Gobain Life Sciences™
082-188-2S
8 mm 1106 psi 457.2 m 1.6 mm
1410.00 €
457m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15610695
View Documents
Saint Gobain Life Sciences™
082-188-3
9.5 mm 1106 psi 15.2 m 2.4 mm
78.55 €
15m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15640695
View Documents
Saint Gobain Life Sciences™
082-250-2
9.5 mm 1106 psi 15.2 m 1.6 mm
66.15 €
15m
In Stock
to see stock.
Edge
Add to basket
15670695
View Documents
Saint Gobain Life Sciences™
082-250-3
11.2 mm 1106 psi 15.2 m 2.4 mm
159.00 €
15m
In Stock
to see stock.
Edge
Add to basket
15680695
View Documents
Saint Gobain Life Sciences™
082-250-3M
11.2 mm 1106 psi 91.4 m 2.4 mm
585.00 €
91m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15690695
View Documents
Saint Gobain Life Sciences™
082-250-3S
11.2 mm 1106 psi 213.3 m 2.4 mm
1305.00 €
213m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15660695
View Documents
Saint Gobain Life Sciences™
082-250-2S
9.5 mm 1106 psi 304.8 m 1.6 mm
1125.00 €
305m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15650695
View Documents
Saint Gobain Life Sciences™
082-250-2M
9.5 mm 1106 psi 106.7 m 1.6 mm
394.00 €
107m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
17923571
View Documents
Saint Gobain Life Sciences™
082-375-2M
12.7 mm 12 psi (at 19°C) 61 m 1.6 mm
376.00 €
61m
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15660705
GREENER_CHOICE
View Documents
Saint Gobain Life Sciences™
082-375-2
12.7 mm 1106 psi 15.2 m 1.6 mm
102.00 €
15m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15630705
View Documents
Saint Gobain Life Sciences™
082-313-3
12.7 mm 1106 psi 15.2 m 2.4 mm N/A N/A N/A
15600705
View Documents
Saint Gobain Life Sciences™
082-250-4
12.7 mm 1106 psi 15.2 m 3.2 mm
134.00 €
15m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15670705
View Documents
Saint Gobain Life Sciences™
082-375-2M
12.7 mm 1106 psi 61.0 m 1.6 mm N/A N/A N/A
15610705
View Documents
Saint Gobain Life Sciences™
082-250-4M
12.7 mm 1106 psi 61.0 m 3.2 mm N/A N/A N/A
15650705
View Documents
Saint Gobain Life Sciences™
082-313-3S
12.7 mm 1106 psi 152.4 m 2.4 mm N/A N/A N/A
15620705
View Documents
Saint Gobain Life Sciences™
082-250-4S
12.7 mm 1106 psi 152.4 m 3.2 mm N/A N/A N/A
17933511
View Documents
Saint Gobain Life Sciences™
082-375-3M
14.3 mm 16 psi (at 19°C) 61 m 2.4 mm
497.00 €
61m
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
17983511
View Documents
Saint Gobain Life Sciences™
082-375-3S
14.3 mm 16 psi (at 19°C) 122 m 2.4 mm
990.00 €
122m
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15600715
View Documents
Saint Gobain Life Sciences™
082-375-3M
14.3 mm 1106 psi 61.0 m 2.4 mm N/A N/A N/A
15620715
View Documents
Saint Gobain Life Sciences™
082-375-4
15.9 mm 1106 psi 15.2 m 3.2 mm
187.00 €
15m
In Stock
to see stock.
Add to basket
15630715
View Documents
Saint Gobain Life Sciences™
082-375-4M
15.9 mm 1106 psi 45.7 m 3.2 mm
523.00 €
46m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15640715
View Documents
Saint Gobain Life Sciences™
082-375-4S
15.9 mm 1106 psi 106.7 m 3.2 mm
1150.00 €
107m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15650715
View Documents
Saint Gobain Life Sciences™
082-500-3
17.4 mm 1106 psi 15.2 m 2.4 mm
172.00 €
15m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15680715
View Documents
Saint Gobain Life Sciences™
082-500-4
19.1 mm 1106 psi 15.2 m 3.2 mm
237.00 €
15m
In Stock
to see stock.
Edge
Add to basket
15690715
View Documents
Saint Gobain Life Sciences™
082-500-4M
19.1 mm 1106 psi 27.4 m 3.2 mm
391.00 €
27m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
17943421
View Documents
Saint Gobain Life Sciences™
082-500-4S
19.1 mm 15 psi (at 19°C) 76 m 3.2 mm
1050.00 €
76m
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15600725
View Documents
Saint Gobain Life Sciences™
082-500-4S
19.1 mm 1106 psi 76.2 m 3.2 mm N/A N/A N/A
15610725
View Documents
Saint Gobain Life Sciences™
082-625-4
22.2 mm 1106 psi 15.2 m 3.2 mm
284.00 €
15m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
15620725
View Documents
Saint Gobain Life Sciences™
082-750-4
25.4 mm 1106 psi 4.5 m 3.2 mm
114.00 €
4.50m
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15630725
GREENER_CHOICE
View Documents
Saint Gobain Life Sciences™
082-750-6
28.3 mm 1106 psi 4.5 m 4.8 mm
172.00 €
4.50m
In Stock
to see stock.
Edge
Add to basket
15630685
GREENER_CHOICE
View Documents
Saint Gobain Life Sciences™
082-1000-6
34.9 mm 1106 psi 4.5 m 4.8 mm
325.00 €
4.50m
In Stock
to see stock.
Edge
Add to basket
15640685
GREENER_CHOICE
View Documents
Saint Gobain Life Sciences™
082-1000-8
38.1 mm 1106 psi 4.5 m 6.4 mm
336.00 €
4.50m
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
Description
Description
C-Flex™ biopharmaceutical tubing is the original patented thermoplastic elastomer tubing specifically designed to meet the critical demands of pharmaceutical and biopharmaceutical applications for fluid processing. C-Flex tubing for biopharma manufacturing is specifically designed for aseptic welding connections and sealing disconnection. C-Flex tubing is manufactured from pharmaceutical-grade thermoplastic elastomer, and full validation and extractable profile testing reports are available to assist in the validation of C-Flex applications.
  • Sealable and weldable either pre- or post- sterilization
  • Sterilizable by gamma and x-ray irradiation as well as autoclave
  • Complies with USP Class VI and ISO standards
  • Full Validation Package available for all promoted C-Flex products
  • Moldable, bondable, and formable for single-use assemblies and overmolds
  • Remains flexible from -45°C to +135°C (-50°F to +275°F)
  • Significantly less permeable than silicone
  • All promoted C-Flex products are Animal-Derived Component-Free (ADCF)
  • C-Flex 072 has excellent durability in peristaltic pump applications
Typical Applications
  • Aseptic Sealing Disconnections
  • Aseptic Welding Connections
  • Fluid Transfer in Pharmaceutical, Biopharmaceutical, and/or Laboratory Settings
  • Single-Use Assemblies
Specifications
Specifications
Autoclavable
Thermoplastic
Tubing
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Saint Gobain Life Sciences™ C-Flex™ 082 TPE Tubing