Learn More
Saint Gobain Life Sciences C-Flex™ 374 TPE Tubing
Biopharmaceutical translucent C-Flex™ 374 TPE Tubing is specifically designed to meet the critical demands of pharmaceutical and biopharmaceutical applications for fluid processing.
18.28 € - 2375.00 €
Specifications
Certifications/Compliance | USP Class VI, ISO 10993, EP |
---|---|
Autoclavable | Autoclavable |
Material | C-Flex |
Elongation | 915% |
For Use With (Application) | Pharmaceutical and Biopharmaceutical Applications, Single-use Assemblies |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Overall Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Overall Length (English) | Price | Quantity & Availability | |||||
15670725
|
Saint Gobain Life Sciences
374-125-2 |
1/8 in. | 1/4 in. | 50 ft. |
89.70 €
15m |
In Stock
Log in to see stock. |
|||||
15630735
|
Saint Gobain Life Sciences
374-188-3 |
3/16 in. | 0.375 in. | 50 ft. |
87.60 €
15m |
In Stock
Log in to see stock. |
|||||
13499698
|
Saint Gobain Life Sciences
374-250-2 |
1/4 in. | 0.375 in. | 50 ft. | N/A | ||||||
15620745
|
Saint Gobain Life Sciences
374-250-4 |
1/4 in. | 0.5 in. | 50 ft. |
169.00 €
15m |
In Stock
Log in to see stock. |
|||||
15650745
|
Saint Gobain Life Sciences
374-313-3 |
5/16 in. | 0.5 in. | 50 ft. |
122.00 €
15m |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15680745
|
Saint Gobain Life Sciences
374-375-2 |
0.375 in. | 0.5 in. | 50 ft. |
347.00 €
15m |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15670755
|
Saint Gobain Life Sciences
374-500-3 |
0.5 in. | 11/16 in. | 50 ft. |
181.00 €
15m |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
15630765
|
Saint Gobain Life Sciences
374-625-4 |
0.625 in. | 0.875 in. | 50 ft. |
256.00 €
15m |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
15640765
|
Saint Gobain Life Sciences
374-750-4 |
0.75 in. | 1 in. | 15 ft. |
118.00 €
4.50m |
In Stock
Log in to see stock. |
|||||
15650765
|
Saint Gobain Life Sciences
374-750-6 |
0.75 in. | 1.125 in. | 15 ft. |
172.00 €
4.50m |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
15650725
|
Saint Gobain Life Sciences
374-1000-6 |
1 in. | 1.375 in. | 15 ft. |
269.00 €
4.50m |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15660725
|
Saint Gobain Life Sciences
374-1000-8 |
1 in. | 1.5 in. | 15 ft. |
336.00 €
4.50m |
In Stock
Log in to see stock. |
|||||
15690725
|
Saint Gobain Life Sciences
374-125-2S |
1/8 in. | 1/4 in. | 2500 ft. |
2375.00 €
762m |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
15620735
|
Saint Gobain Life Sciences
374-188-2S |
3/16 in. | 5/16 in. | 1500 ft. |
1410.00 €
457m |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
15680735
|
Saint Gobain Life Sciences
374-250-2S |
1/4 in. | 0.375 in. | 1000 ft. |
1125.00 €
305m |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
15610745
|
Saint Gobain Life Sciences
374-250-3S |
1/4 in. | 7/16 in. | 700 ft. |
1305.00 €
213m |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
15640745
|
Saint Gobain Life Sciences
374-250-4S |
1/4 in. | 0.5 in. | 500 ft. |
1160.00 €
152m |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
15670745
|
Saint Gobain Life Sciences
374-313-3S |
5/16 in. | 0.5 in. | 500 ft. | N/A | ||||||
15660755
|
Saint Gobain Life Sciences
374-375-4S |
0.375 in. | 0.625 in. | 350 ft. |
1150.00 €
107m |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
15620765
|
Saint Gobain Life Sciences
374-500-4S |
0.5 in. | 0.75 in. | 250 ft. | N/A | ||||||
15680725
|
Saint Gobain Life Sciences
374-125-2M |
1/8 in. | 1/4 in. | 500 ft. |
510.00 €
152m |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
15610735
|
Saint Gobain Life Sciences
374-188-2M |
3/16 in. | 5/16 in. | 350 ft. | N/A | ||||||
15670735
|
Saint Gobain Life Sciences
374-250-2M |
1/4 in. | 0.375 in. | 350 ft. |
414.00 €
107m |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
15600745
|
Saint Gobain Life Sciences
374-250-3M |
1/4 in. | 7/16 in. | 300 ft. |
892.00 €
91m |
In Stock
Log in to see stock. |
|||||
15630745
|
Saint Gobain Life Sciences
374-250-4M |
1/4 in. | 0.5 in. | 200 ft. | N/A | ||||||
15690745
|
Saint Gobain Life Sciences
374-375-2M |
0.375 in. | 0.5 in. | 200 ft. | N/A | ||||||
15620755
|
Saint Gobain Life Sciences
374-375-3M |
0.375 in. | 0.563 in. | 200 ft. | N/A | ||||||
15650755
|
Saint Gobain Life Sciences
374-375-4M |
0.375 in. | 0.625 in. | 150 ft. |
611.00 €
46m |
Estimated Shipment: 30-05-2025
Log in to see stock. |
|||||
15610765
|
Saint Gobain Life Sciences
374-500-4M |
0.5 in. | 0.75 in. | 90 ft. |
391.00 €
27m |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
Description
C-Flex™ biopharmaceutical tubing is the original patented thermoplastic elastomer tubing specifically designed to meet the critical demands of pharmaceutical and biopharmaceutical applications for fluid processing. C-Flex tubing for biopharma manufacturing is specifically designed for aseptic welding connections and sealing disconnection. C-Flex tubing is manufactured from pharmaceutical-grade thermoplastic elastomer, and full validation and extractable profile testing reports are available to assist in the validation of C-Flex applications.
- Sealable and weldable either pre- or post- sterilization
- Sterilizable by gamma and x-ray irradiation as well as autoclave
- Complies with USP Class VI and ISO standards
- Full Validation Package available for all promoted C-Flex products
- Moldable, bondable, and formable for single-use assemblies and overmolds
- Remains flexible from -45°C to 135°C (-50°F to 275°F)
- Significantly less permeable than silicone
- All promoted C-Flex products are Animal-Derived Component-Free (ADCF)
- C-Flex 072 has excellent durability in peristaltic pump applications
Typical Applications
- Aseptic Sealing Disconnections
- Aseptic Welding Connections
- Fluid Transfer in Pharmaceutical, Biopharmaceutical, and/or Laboratory Settings
- Single-Use Assemblies
Specifications
USP Class VI, ISO 10993, EP | |
C-Flex | |
Pharmaceutical and Biopharmaceutical Applications, Single-use Assemblies | |
Non-sterile | |
60 (Shore A) | |
Tubing |
Autoclavable | |
915% | |
-45°C | |
Translucent | |
Flexible |