missing translation for 'onlineSavingsMsg'
Learn More

IKA™ C-MAG MS 10 Magnetic Stirrer
GREENER_CHOICE

Magnetic stirrer without heating features a ceramic set-up plate, which offers excellent chemical resistance.

Brand:  IKA™ 0003582602

851.24 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 16298141

  • 965.00 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Powerful motor for stirring quantities of up to 15 L (H2O)
  • Elevated control panel for protection against leaking liquids
  • Speed range: 100 - 1500 rpm
Specifications

Specifications

100 to 1500 rpm
230 V
415 x 330 x 105 mm
260 mm
UK-Style
Ceramic
30 W
260 mm
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.