Learn More
Thermo Scientific Chemicals C-Myc Peptide
For cellular and molecular biology applications
Brand:  Thermo Scientific Chemicals J66846.M^
 View more versions of this product
Product Code. 15458369
  • 49.45 € / 1mg
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HPLC
Solid
C-Myc Peptide
RUO - research use only
-30°C to -10°C
1203.29
1 mg
Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu