Learn More
IKA™ C1 Calorimeter Package with C1.12 Combustion Chamber
The C1 Calorimeter Package with C1.20 Combustion Chamber combines a compact design with a high degree of automation, featuring a light, halogen-resistant combustion chamber.
Brand: IKA™ 0010005638
View more versions of this product
This item is not returnable.
View return policy
Description
- Features a high degree of automation
- Compact design offers a small footprint
- Operates in accordance with DIN 51900 and ISO 1928
- PCI / PCS measurement analysis follows classic isoperibolic methodology
- Easily retractable combustion chamber with active catalytic coating and halogen-resistance is specially designed for the combustion of sample materials containing halogens and sulfur (>3%)
- Compatible with IKA™ C6040 Calwin calorimeter software (sold separately), which allows additional extensions and adaptations, such as the management of measurement data and LIMS
- Includes wear parts and consumables for the first 500 tests including 25 calibrations
- C1.10 combustion chamber is fitted with a large C5010.5 crucible support and a large C6 quartz crucible
Specifications
Calorimeter | |
50/60 Hz | |
Up to 40,000 J | |
100/240 V |
DIN 51900, ISO 1928 | |
C1 Calorimeter, C1.12 Halogen-resistant Combustion Chamber, C1.20 Heater | |
20°C to 30°C |
Product Content Correction
Product Title
IKA™ C1 Calorimeter Package with C1.12 Combustion Chamber > C1/C1.12 Calorimeter Package; Includes: C1 Calorimeter, C1.12 Halogen-resistant Combustion Chamber, C1.20 Heater
Spot an opportunity for improvement?Share a Content Correction