missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf106 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | C14orf106 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16864775
|
Proteintech
26264-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16854775
|
Proteintech
26264-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIS18BP1 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.Specifications
C14orf106 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6P0N0 | |
55320 | |
C14orf106 Fusion Protein Ag23483 | |
Primary | |
-20°C | |
MIS18BP1 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C14orf106, HSA242977, HsKNL 2, KIAA1903, KNL2, M18BP1, Mis18 binding protein 1, MIS18BP1, P243 | |
MIS18BP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title