Learn More
Tocris Bioscience™ C25-140
TRAF6-Ubc13 interaction inhibitor
Brand:  Tocris Bioscience™ 7715/10
249.67 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18766926
  • 316.00 € / 10mg
Explore more special offers
Description
Description
C25-140 binds directly to TNF receptor-associated factor 6 (TRAF6) and inhibits its interaction with ubiquitin-conjugating enzyme Ubc13. C25-140 reduces TRAF6-Ubc13 activity and decreases NF-κB activation and inflammatory signaling in vitro. C25-140 exhibits selectivity for TRAF6 over other E3 ligases except cIAP1. In in vivo mouse models of autoimmune psoriasis and rheumatoid arthritis, the compound improves disease symptoms. The compound also suppresses the pro-inflammatory signaling activity of IL-17A and prevents its inhibitory effect on the anti-HBV action of IFN-α HepG2 cells.
Specifications
Specifications
3-[3,5-Dimethyl-1-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)-1H-pyrazol-4-yl]-1-[4-(phenylmethyl)-1-piperidinyl]-1-propanone
10 mg
C26H31N7O
1358099-18-9
Ubiquitin Ligase (E3) Inhibitors