missing translation for 'onlineSavingsMsg'
Learn More

uvex C500 Cut Protection Gloves

Product Code. 17200431
Click to view available options
Size:
6
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Uvex C500 gloves combine comfort, flexibility, and tactility with the highest level of cut protection (level 5/C). Soft-grip coating provides mechanical abrasion resistance.

  • Bamboo TwinFlex™ Technology ensures a very high level of cut protection
  • Soft-grip coating provides mechanical abrasion resistance
  • Highly flexible with good tactile sensation
  • Uvex Climazone effectively removes excess warmth and moisture
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Silicone-free according to the imprint test
  • 3D Ergo Technology ensures a secure fit
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Uncoated
Color Green
Material Bamboo Viscose, HPPE, Polyamide
Size 6
EN388 Standard 1X4XC
For Use With (Application) Automotive, Construction, Glass Handling, Logistics, Maintenance, and Sheet Metal Processing
Gender Unisex
Glove Cuff Style Knit
Series C500
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.