Learn More
Labconco™ Cable Clamp
Labconco™ Cable Clamp is a replacement clamp designed for use with sash cables in Protector™ Fume Hoods.
Brand:  Labconco™ 1920100
Product Code. 15804868
  • 3.45 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement clamp for use with Sash cables
  • For use with a variety of Protector™ Laboratory Fume Hoods, including:
    • Classmate™
    • PVC Perchloric Acid and Acid Digestion
    • Stainless Steel Radioisotope
    • Xstream
    • XL Floor Mounted
Specifications
Specifications
Protector™ fume hoods
Cable Sash Clamp