Learn More
Cacodylic acid sodium salt trihydrate 98+%, Thermo Scientific Chemicals
Description
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.

Specifications
Specifications
Linear Formula | (CH3)2AsO2Na·3H2O |
Quantity | 25 g |
UN Number | UN1688 |
Beilstein | 3702348 |
Merck Index | 14,8595 |
Solubility Information | Soluble in water and alcohol. Insoluble in diethyl ether. |
Formula Weight | 214.03 (159.98 Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | Cacodylic acid sodium salt trihydrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10205571 | Certificate of Analysis | 23/11/2021 | A18139.22,A18139.14,A18139.36 |
10237280 | Certificate of Analysis | 23/11/2021 | A18139.22,A18139.14,A18139.36 |
10200826 | Certificate of Analysis | 23/11/2021 | A18139.22,A18139.14,A18139.36 |
10227783 | Certificate of Analysis | 23/11/2021 | A18139.22,A18139.14,A18139.36 |
10211941 | Certificate of Analysis | 23/11/2021 | A18139.22,A18139.14,A18139.36 |
Safety and Handling



- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 2
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.