Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
Brand:  Cadence Science™ 9923
 View more versions of this product
Product Code. 15282035
  • 695.00 € / Pack of 100
Explore more special offers
Description
Description
  • Nontoxic, nonpyrogenic
  • PTFE tubing is flexible and softer than other types of tubing
  • Low friction protects delicate organs
  • Translucent for easy confirmation of reagent delivery
  • Plastic luer hubs; soft silicone tips
  • Sterile, packaged in pouches
  • Five needles per pouch, 20 pouches per box
Specifications
Specifications
Soft, Flexible Animal Feeding Needle
Single-use
18G
2 in.
0.078 in.
PTFE
For critical applications requiring sterile, flexible, single use feeding needles
Sterile
0.5 cm
0.02 cm