Learn More
Cadence Science™ Flexible Plastic (PTFE) Needles with Bite Protectors
Covered at the hub end with 304 stainless steel tubing – protects tubing from damage due to biting during use.
Brand:  Cadence Science™ 9928B
 View more versions of this product
Product Code. 17469373
  • 682.00 € / Pack of 100
Explore more special offers
Specifications
Single-use
18G
1.5 in.
Animal feeding
Sterile
2 mm