missing translation for 'onlineSavingsMsg'
Learn More

Cadence Science™ Reusable Small Animal Feeding Needles: Curved

Precision-assembled stainless-steel feeding needles ideal for feeding and dosing

Brand:  Cadence Science™ 7917

 View more versions of this product

Product Code. 17247470

  • 920.00 € / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Precision-assembled stainless-steel feeding needles ideal for feeding and dosing

  • For the intubation of laboratory animals
  • Straight, curved tubing styles
  • Many standard sizes available, from small to large
  • Curved
  • Autoclavable

Small Animal Needles

  • For animals 0.7 oz. to 1 lb., 5 oz. (20 to 600g)

Specifications

Specifications

Autoclavable
Stainless Steel
Small animal, ideal for feeding and dosing
3 in.
0.157 in.
Curved Animal Feeding Needle
Reusable
14G
0.76 cm
0.04 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.