missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate dihydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11351938
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00012449
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 266.54 (230.50 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.01
Chemical Name or Material Cadmium acetate dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'