Learn More
Cadmium arsenide, 99% (metals basis), Thermo Scientific Chemicals
163.82 €
Chemical Identifiers
CAS | 12006-15-4 |
---|---|
Molecular Formula | As2Cd3 |
Molecular Weight (g/mol) | 487.085 |
MDL Number | MFCD00049723 |
InChI Key | FSIONULHYUVFFA-UHFFFAOYSA-N |
Synonym | cadmium arsenide |
PubChem CID | 6391215 |
SMILES | [As]=[Cd].[As]=[Cd].[Cd] |
Description
Cadmium arsenide has a electronic properties and is used in 3D crystal structure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12006-15-4 | |
487.085 | |
FSIONULHYUVFFA-UHFFFAOYSA-N | |
6391215 |
As2Cd3 | |
MFCD00049723 | |
cadmium arsenide | |
[As]=[Cd].[As]=[Cd].[Cd] |
Specifications
12006-15-4 | |
Lump | |
Cd3As2 | |
UN1557 | |
Soluble in HNO3(nitric acid). | |
[As]=[Cd].[As]=[Cd].[Cd] | |
6391215 | |
99% | |
3.031 g/mL | |
Cadmium arsenide |
Gray | |
As2Cd3 | |
MFCD00049723 | |
cadmium arsenide | |
FSIONULHYUVFFA-UHFFFAOYSA-N | |
487.085 | |
487.04 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H300-H330-H350
Fatal if swallowed.
Fatal if inhaled.
May cause cancer.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
H301-H330-H350i
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 234-484-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.