missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromate hydrate, 98%, Thermo Scientific™

Product Code. 11335149
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14518-94-6
Molecular Formula Br2CdO6
Molecular Weight (g/mol) 368.22
MDL Number MFCD04971194
InChI Key VECQEXBULIKSDM-UHFFFAOYSA-L
Synonym cadmium bromate, cadmium dibromate, cadmium 2+ dibromate, 2bro3.cd, acmc-1c8r8, bromic acid, cadmiumsalt 8ci,9ci
PubChem CID 14598111
SMILES [Cd++].[O-][Br](=O)=O.[O-][Br](=O)=O

Specifications

Quantity 25 g
Linear Formula Cd(BrO3)2·xH2O
UN Number UN1450
IUPAC Name cadmium(2+);dibromate
Formula Weight 368.21 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Cadmium bromate hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'