missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11333118
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13464-92-1
Molecular Formula Br2CdH8O4
Molecular Weight (g/mol) 344.282
MDL Number MFCD00150204
InChI Key PANYFCXNZSEXSU-UHFFFAOYSA-L
Synonym cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent
PubChem CID 3084147
IUPAC Name cadmium(2+);dibromide;tetrahydrate
SMILES O.O.O.O.[Br-].[Br-].[Cd+2]

Specifications

Physical Form Powder
Quantity 100 g
UN Number UN2570
Solubility Information Soluble in water.
Formula Weight 344.28 (272.22 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Cadmium bromide tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'